Z290564
Właściwości
aluminum housing
pkg of 1 ea
cell culture: suitable
47 mm
Opis
Pokaż różnice
1 of 1
Sigma-Aldrich
Vacuum filtration assembly replacement parts
Z290777
Z290572
Z290459
Vacuum filtration assembly flasks
aluminum clamp for 47mm assemblies
PTFE gasket
#8 silicone stopper
side-arm flask, capacity 1000 mL
-
pkg of 1 ea, pack of 1
silicone housing
borosilicate glass
41 mm
Explore all of our products under Vacuum filtration assembly replacement parts
Wybierz jedną z najnowszych wersji:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.
Odwiedź Bibliotekę dokumentów
Powiązane kategorie
Cell Culture & Cryogenics
Cell Culture Supplies
Filtration
Labware
Vacuum Filters