Przejdź do
Z290505
Sigma-Aldrich® vacuum filtration assembly
for 47 mm filters with stainless steel screen, #8 stopper, same as Z290432 but without side arm flask
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1470,00 zł
Informacje o tej pozycji
NACRES:
NB.24
UNSPSC Code:
41104900
1470,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
material
glass housing, stainless steel membrane
feature
cap plug seal, graduation
packaging
pkg of 1 ea
neck diam.
41 mm
General description
Same as Z290432 but without side arm flask.
Vacuum filtration apparatus for HPLC solvent purification, particulate analysis, bacteriological studies, radiochemical determinations, quantitative gravimetric analysis, and similar applications. Clamp seals filter membrane between flanges on funnel top and funnel stem.
Includes: Funnel top, funnel stem/membrane support, aluminum clamp, stopper (except standard taper units) and filtration flask (where indicated).
For replacement parts, visit our online glassware catalog at www.sigma-aldrich.com
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z290440Z290475Z290416
description
for 47 mm filters with stainless steel screen, #8 stopper, same as Z290432 but without side arm flask
description
for 47 mm filters with glass support, #8 stopper, same as Z290432 but without filtration flask
description
for 25 mm filters with glass support, #5 stopper, same as Z290467 but without filtration flask
description
for 47 mm filters with glass support, NS 40/35 joints, 4 L flask (included)
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
pkg of 1 ea
packaging
-
material
stainless steel membrane, glass housing
material
glass membrane, glass housing
material
glass housing, glass membrane
material
-
feature
graduation, cap plug seal
feature
graduation
feature
graduation, cap plug seal
feature
-
neck diam.
41 mm
neck diam.
41 mm
neck diam.
27 mm
neck diam.
-
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)