Merck
T3156
Microcentrifuge tubes, extended capacity
volume 1.7 mL , ungraduated, non-siliconized polypropylene
Wybierz wielkość
Informacje o tej pozycji
NACRES:
NB.11
UNSPSC Code:
41121703
Pozwól nam pomóc
material
cap, non-siliconized polypropylene
sterility
non-sterile
packaging
pack of 500
volume
1.7 mL , ungraduated
General description
The tubes listed below are specifically designed to hold more volume than other comparable tubes while still fitting all of the most commonly used microcentrifuges. Writing surface on flat lids and side wall for quick sample ID.
The lids provide an easily punctured syringe port for easy access to the contents without opening the tube. Siliconized tubes simplify research requiring low adhesion surface.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)