Przejdź do
HS120098
Pearl Pipette
adjustable, volume 0.5-10 μL, light blue, single-channel
Wybierz wielkość
Zmień widok
Do Państwa/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
931,00 zł
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Material:
light blue , white
931,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
Pomoc techniczna
Pozwól nam pomóc
material
light blue , white
feature
adjustable, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific HS120098
volume measuring range
0.5-10 μL
General description
The Pearl is a single-channel pipette offering adjustable volume and precision liquid handling. The sleek, ergonomic design and reduced plunger pressure help reduce the chance of Repetitive Stress Injury (RSI), as the pipette requires 2/3 the operation pressure of most standard pipettes. The Pearl is tested in accordance with standards and a calibration certificate is supplied. Manufactured in accordance with ISO08655 and GLP regulations.
Features and Benefits
  • Internal metal parts for increased accuracy and calibration stability, and long life
  • Ergonomic Slight downward slant to tip ejector button reduces hand fatigue, incidence of repetitive strain injury (RSI)
  • Easy volume adjustment with locking tab to prevent accidental change
  • Separate tip ejector
  • Large digit volume display
  • Color coded and labeled with volume range
  • Both top and side have colored coded components for quick visual confirmation
  • CE certified and Manufactured in accordance with ISO08655 and GLP regulations
  • Autoclavable
Legal Information
Pearl is a trademark of Specac, Inc.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
HS120101HS120104HS120218
description
adjustable, volume 0.5-10 μL, light blue, single-channel
description
adjustable, volume 10-100 μL, blue, single-channel
description
adjustable, volume 100-1000 μL, blue, single-channel
description
adjustable, volume 30-300 μL, blue, 12-channel
material
light blue , white
material
blue , white
material
blue , white
material
blue , white
volume measuring range
0.5-10 μL
volume measuring range
10-100 μL
volume measuring range
100-1000 μL
volume measuring range
30-300 μL
manufacturer/tradename
Heathrow Scientific HS120098
manufacturer/tradename
Heathrow Scientific HS120101
manufacturer/tradename
Heathrow Scientific HS120104
manufacturer/tradename
Heathrow Scientific HS120218
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
feature
adjustable
feature
adjustable, single-channel
feature
adjustable, single-channel
feature
12-channel, adjustable
Still not finding the right product?
Explore all of our products under Pearl Pipette
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)