Merck
H1784
Modular hybridization system
Adhesive module A, 0.6 mm depth
About This Item
Kod UNSPSC:
41100000
Kompatybilność
Reusable, press-to-seal gasket and cover slip enclosures which can be quickly assembled and disassembled for hybridizing and imaging thick or free floating specimens of any thickness with up to a 13 mm diameter. Build up to three chambers per slide. Minimize specimen handling, eliminate evaporation and leaking even when submerged. RNase-free, ready to use for multiple incubations and agitated washing steps. (Assortment pack contains seal tabs, 5 ea A-D & 10 each E).
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)