Merck
F8395
Carestream® electron microscopy film 4489
Wybierz wielkość
Informacje o tej pozycji
NACRES:
MA.01
UNSPSC Code:
41105300
Pozwól nam pomóc
sheet size
3 1/4 in. × 4 in.
storage temp.
2-8°C
General description
Fine-grain, blue-sensitive emulsion on 7-mil ESTAR base, notched for emulsion identification. Tear- and abrasion-resistant, dimensionally stable. Requires shorter pump-down time and curls less than Kodak electron image film SO-163.
Legal Information
Kodak is a registered trademark of Eastman Kodak Company
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)