Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
100 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 3760,00 zł |
Informacje o tej pozycji
3760,00 zł
material
polypropylene , polypropylene plate, round bottom clear , translucent bottom
sterility
non-sterile
feature
lid: no, skirt
manufacturer/tradename
Corning 3959
packaging
bag of 5 ×
size
96 wells
well volume
1 mL
wells
96 wells , storage block
suitability
suitable for (cell cuture)
binding type
non-treated surface
General description
All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 96 well polypropylene plates are built to the industry standard footprint. The plates are available in round, flat, and V-bottom formats to meet a wide variety of assay and storage needs.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description round bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no | description V-bottom clear, polypropylene, bag of 10 ×, non-sterile, lid: no | description V-bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no | description round bottom clear, polypropylene, bag of 5 ×, sterile, lid: no |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| size 96 wells | size 96 wells | size 96 wells | size 96 wells |
| material polypropylene plate, round bottom clear , translucent bottom | material polypropylene plate, translucent bottom, V-bottom clear | material polypropylene plate, V-bottom clear , translucent bottom | material translucent bottom, polypropylene , round bottom clear , polypropylene plate |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility sterile |
| manufacturer/tradename Corning 3959 | manufacturer/tradename Corning 3957 | manufacturer/tradename Corning 3961 | manufacturer/tradename Corning 3958 |
| packaging bag of 5 × | packaging bag of 10 × | packaging bag of 5 × | packaging bag of 5 × |
Still not finding the right product?
Explore all of our products under Corning® 96 well Polypropylene Deep Well Plate
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?