Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
100 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 4370,00 zł |
Informacje o tej pozycji
4370,00 zł
material
V-bottom clear , polypropylene , polypropylene plate, translucent bottom
sterility
non-sterile
feature
lid: no, skirt
manufacturer/tradename
Corning 3957
packaging
bag of 10 ×
size
96 wells
well volume
0.5 mL
wells
96 wells , storage block
suitability
suitable for (cell cuture)
binding type
non-treated surface
General description
All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 96 well polypropylene plates are built to the industry standard footprint. The plates are available in round, flat, and V-bottom formats to meet a wide variety of assay and storage needs.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description V-bottom clear, polypropylene, bag of 10 ×, non-sterile, lid: no | description V-bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no | description V-bottom clear, polypropylene, bag of 5 ×, sterile, lid: no | description round bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no |
| size 96 wells | size 96 wells | size 96 wells | size 96 wells |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| material translucent bottom, V-bottom clear , polypropylene plate | material translucent bottom, polypropylene plate, V-bottom clear | material polypropylene , V-bottom clear , translucent bottom, polypropylene plate | material polypropylene , translucent bottom, polypropylene plate, round bottom clear |
| feature skirt, lid: no | feature skirt, lid: no | feature lid: no, skirt | feature skirt, lid: no |
| well volume 0.5 mL | well volume 2 mL | well volume 2 mL | well volume 1 mL |
| binding type non-treated surface | binding type non-treated surface | binding type non-treated surface | binding type non-treated surface |
Still not finding the right product?
Explore all of our products under Corning® 96 well Polypropylene Deep Well Plate
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?