Wybierz wielkość
| Do Państwa/SKU | Dostępność | Cena netto |
|---|---|---|
25 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 1120,00 zł |
Informacje o tej pozycji
1120,00 zł
material
polypropylene , polypropylene plate, round bottom clear , translucent bottom
sterility
sterile
feature
lid: no, skirt
manufacturer/tradename
Corning 3958
packaging
bag of 5 ×
size
96 wells
well volume
1 mL
wells
96 wells , storage block
suitability
suitable for (cell cuture)
binding type
non-treated surface
General description
All plates and blocks are made from polypropylene for solvent resistance and are ideal for compound library, combinatorial chemistry and genomics applications. The 96 well polypropylene plates are built to the industry standard footprint. The plates are available in round, flat, and V-bottom formats to meet a wide variety of assay and storage needs.
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description round bottom clear, polypropylene, bag of 5 ×, sterile, lid: no | description V-bottom clear, polypropylene, bag of 5 ×, sterile, lid: no | description round bottom clear, polypropylene, bag of 5 ×, non-sterile, lid: no | description V-bottom clear, polypropylene, bag of 10 ×, sterile, lid: no |
| size 96 wells | size 96 wells | size 96 wells | size 96 wells |
| suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) | suitability suitable for (cell cuture) |
| material polypropylene , round bottom clear , polypropylene plate, translucent bottom | material V-bottom clear , polypropylene , polypropylene plate, translucent bottom | material polypropylene , polypropylene plate, round bottom clear , translucent bottom | material V-bottom clear , polypropylene , polypropylene plate, translucent bottom |
| sterility sterile | sterility sterile | sterility non-sterile | sterility sterile |
| well volume 1 mL | well volume 2 mL | well volume 1 mL | well volume 0.5 mL |
| packaging bag of 5 × | packaging bag of 5 × | packaging bag of 5 × | packaging bag of 10 × |
Still not finding the right product?
Explore all of our products under Corning® 96 well Polypropylene Deep Well Plate
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?