Merck
BR29953
BRAND® BLAUBRAND® bulb pipette with serialized numbers, calibrated to deliver (TD, EX)
capacity 2 mL, one mark
Wybierz wielkość
Informacje o tej pozycji
UNSPSC Code:
41121510
Capacity:
2 mL
Material:
AR-Glass
Feature:
supplementary to DIN range
Packaging:
pkg of 12 ea
Pozwól nam pomóc
material
AR-Glass
feature
supplementary to DIN range
packaging
pkg of 12 ea
manufacturer/tradename
BRAND 27452
capacity
2 mL
General description
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.According to DIN EN ISO 648.
Legal Information
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)