Merck
A2171
N-(6-Aminohexyl)-2-naphthalenesulfonamide hydrochloride
Informacje o tej pozycji
UNSPSC Code:
12352200
Pomoc techniczna
Pozwól nam pomóc
storage temp.
2-8°C
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
H Hidaka et al.
Proceedings of the National Academy of Sciences of the United States of America, 78(7), 4354-4357 (1981-07-01)
N-(6-Aminohexyl)-5-chloro-1-naphthalenesulfonamide (W-7) and its derivatives are putative calmodulin antagonists that bind to calmodulin and inhibit Ca2+/calmodulin-regulated enzyme activities. Autoradiographic studies using tritiated W-7 showed that this compound penetrates the cell membrane, is distributed mainly in the cytoplasm, and inhibits proliferation