Merck
89469
LL-2,6-Diaminopimelic acid
Synonim(y):
(2S,6S)-2,6-Diaminoheptanedioic acid, L,L-α,ε-Diaminopimelic acid, L-threo-2,6-Diaminopimelic acid, L-DAP
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C7H14N2O4
Numer CAS:
Masa cząsteczkowa:
190.20
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Beilstein/REAXYS Number:
1726901
Assay:
≥95.0% (TLC)
Form:
powder
Pozwól nam pomóc
InChI
1S/C7H14N2O4/c8-4(6(10)11)2-1-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)/t4-,5-/m0/s1
InChI key
GMKMEZVLHJARHF-WHFBIAKZSA-N
assay
≥95.0% (TLC)
form
powder
optical purity
enantiomeric excess: ≥98.0%
Klasa składowania
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Isolation and characterization of antagonistic actinomycetes from marine soil.
Deepika, L. and Kannabiran, K.
Journal of Microbial & Biochemical Technology, 2, 1-6 (2010)
J H Yoon et al.
International journal of systematic bacteriology, 48 Pt 1, 187-194 (1998-05-02)
The 16S rDNAs from 38 strains of the genus Nocardioides, two Aeromicrobium species and Terrabacter tumescens were directly sequenced and then analysed. The mean nucleotide similarity value between the type strains of validly described Nocardioides species was 96.1 +/- 3.0%.
Chemical composition as a criterion in the classification of aerobic actinomycetes.
Lechevalier, M. P. and Lechevalier, H.
International Journal of Systematic Bacteriology, 20, 435-443 (1970)
Yuchen Liu et al.
Journal of bacteriology, 192(13), 3304-3310 (2010-04-27)
The pathway of lysine biosynthesis in the methanococci has not been identified previously. A variant of the diaminopimelic acid (DAP) pathway uses diaminopimelate aminotransferase (DapL) to catalyze the direct conversion of tetrahydrodipicolinate (THDPA) to ll-DAP. Recently, the enzyme DapL (MTH52)
M A de Pedro et al.
Proceedings of the National Academy of Sciences of the United States of America, 78(9), 5856-5860 (1981-09-01)
In vivo studies of murein biosynthesis show that newly synthesized murein unexpectedly differs in its chemical composition from preexisting murein. New murein is loosely crosslinked with the preexisting sacculus; in a maturation process involving further transpeptidation, the final stage of