Merck
08515
Melamine
pharmaceutical impurity standard, ≥95.0% (HPLC)
Synonim(y):
Metformin impurity D (Ph Eur), 2,4,6-Triamino-1,3,5-triazine, sym-Triaminotriazine
Informacje o tej pozycji
Wzór empiryczny (zapis Hilla):
C3H6N6
Numer CAS:
Masa cząsteczkowa:
126.12
EC Number:
203-615-4
UNSPSC Code:
41116107
PubChem Substance ID:
Beilstein/REAXYS Number:
124341
MDL number:
Przejdź do
Pomoc techniczna
Pozwól nam pomóc
grade
analytical standard
assay
≥95.0% (HPLC)
form
solid
shelf life
limited shelf life, expiry date on the label
impurities
≤10% related substances, ≤5.0% solvents, ≤5.0% water
mp
>300 °C (lit.)
application(s)
pharmaceutical (small molecule)
storage temp.
2-8°C
SMILES string
Nc1nc(N)nc(N)n1
InChI
1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)
InChI key
JDSHMPZPIAZGSV-UHFFFAOYSA-N
pictograms
Health hazard
signalword
Warning
Hazard Classifications
Carc. 2 - Repr. 2 - STOT RE 2
target_organs
Urinary tract
Klasa składowania
11 - Combustible Solids
wgk
WGK 1
flash_point_f
572.0 °F - closed cup
flash_point_c
300 °C - closed cup
Wykazy regulacyjne
Wykazy regulacyjne dotyczą głównie produktów chemicznych. Można w nich podawać ograniczoną liczbę informacji na temat produktów niechemicznych. Brak wpisu oznacza, że żaden ze składników nie znajduje się w wykazie. Użytkownik odpowiada za zagwarantowanie bezpiecznego i zgodnego z prawem stosowania produktu.
108-78-1
CAS No.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)
Odwiedź Bibliotekę dokumentów
Ana Paula Craig et al.
Annual review of food science and technology, 4, 369-380 (2013-01-10)
Surface-enhanced Raman spectroscopy (SERS) is an advanced Raman technique that enhances the vibrational spectrum of molecules adsorbed on or in the vicinity of metal particles and/or surfaces. Because of its readiness, sensitivity, and minimum sample preparation requirements, SERS is being
Hui Yang et al.
Nano letters, 15(3), 1730-1735 (2015-02-11)
We detect by optical microscopy Au and fluorescent nanoparticles (NPs) during their motion in water-based medium, using an array of dielectric microspheres that are patterned in a microwell array template. The microspheres act as lenses focusing the light originating from
D Frösch et al.
Electron microscopy reviews, 2(2), 231-255 (1989-01-01)
Melamine resins are derived from the heterocyclic compound triaminotriazine, C3H6N6. Similar to proteins in structure and reactivity, water-soluble melamine resins can be used as water-embedding media for electron microscopy (Bachhuber and Frösch, 1983). The idea behind this approach was to
Rishikesh P Dalal et al.
Nature reviews. Nephrology, 7(5), 267-274 (2011-03-23)
Several well-documented outbreaks of melamine poisoning have occurred in both animals and humans during the past 7 years, which led to the identification of melamine and cyanuric acid as nephrotoxins. This Review provides an overview of the known experimental and
Lei An et al.
Neurobiology of learning and memory, 100, 18-24 (2012-12-13)
Our previous studies reported that hippocampus was one of the target sites of melamine, by which the spatial cognition and hippocampal long-term potentiation (LTP) could be impaired. The aim of present study was to investigate whether cognitive behavior impairment induced