Kluczowe dokumenty
SYMBA4VP1
Millipore
Steritest® Symbio Pumps Validation Protocol
A4 paper format
About This Item
Kod UNSPSC:
41116500
NACRES:
NB.79
Nie można obecnie kupić produktu SYMBA4VP1. Skontaktuj się z zespołem ds. pomocy technicznej
Poziom jakości
kompatybilność
for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio Pumps
Opis ogólny
Steritest® Symbio Pumps validation protocol is designed to make it easier for the technicians to ensure compliance of the symbio pumps for consistent performance. The product is GMP compliant and includes Validation Master Plan (VMP), Installation Qualification (IQ), Operational Qualification (OQ) and Performance Qualification (PQ) test protocols. In addition to the above, ready to be filled report table formats are also provided.
Zastosowanie
Steritest® Symbio Pumps validation protocol is a ready to use GMP compliant validation protocol along with ready to be filled report table formats.
Informacje prawne
STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany