Kluczowe dokumenty
1.50611
Supelco
CARS Complete System (230V,50/60Hz)
for use with SeQuant anion membrane supressor
About This Item
Kod UNSPSC:
41115700
metody
ion chromatography: suitable
Poziom jakości
kompatybilność
for use with SeQuant anion membrane supressor
Opis ogólny
SeQuant CARS (Continuous Anion Regeneration System) with SAMS (SeQuant Anion Membrane Suppressor) will enhance and simplify your anion chromatography analysis. The continuous chemical suppression of the eluent reduces the background and increases the signal for the analyte anions.
This CARS system requires a SAMS suppressor for operation.
continuous auto-regeneration system for SAMS suppressors SeQuant
Informacje prawne
SeQuant is a trademark of Merck KGaA, Darmstadt, Germany