Merck
Wszystkie zdjęcia(4)
Kluczowe dokumenty
Z635405
Swiss Boy lab jacks
load ≤150 kg (with wheels, middle position), platform L × W 400 mm × 400 mm, Model 122R
About This Item
Kod UNSPSC:
41100000
producent / nazwa handlowa
Grauer
Parametry
≤150 kg load (with wheels, middle position)
253-629 mm vertical limit
platforma dł. × szer.
400 mm × 400 mm
kolor
yellow
Opis ogólny
Jack can be raised with load in place.
These high quality Swiss made jacks have massive cast aluminum frames with a chemically resistant anodised finish. Swiss boy jacks are designed to carry heavy loads safely. Available either in standard size or with extended carrying plates for large pots or pans. Units carrying more that 90kg are also available with a motorized lift adjustment for enhanced convenience. The max load is for the middle position. Supplied with top and bottom plates.
Products with motorized jacks are available in Europe only.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)