Z557854
Sigma-Aldrich® temperature controller for ceramic top hotplate stirrer
About This Item
Kod UNSPSC:
41102406
zakres pomiaru temperatury
accuracy: 1 °C
Opis ogólny
The digital temperature controller has fuzzy logic control which provides constant feedback to hot plate to ensure perfect temperature control of the medium without overshooting the set temperature. Connects to the stirring hot plates with a DIN 12878 socket. Includes a 250 mm long × 3 mm diameter stainless steel temperature probe.
Informacje prawne
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
produkt podobny
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)