Z507172
Michel-Miller Swagelok® adapter
Nylon, NPTF 1/8 in., Ace-Thred for #15
About This Item
Kod UNSPSC:
12352200
NACRES:
SB.52
Materiały
Nylon
producent / nazwa handlowa
Ace Glass 584420
metody
LPLC: suitable
gwint wewn.
1/8 in.
dopasowywanie
Ace-Thred for #15
Opis ogólny
Use to make Swagelok tubing connections with Ace threaded glassware. One end is Ace-Thred, other is NPTF. Connects tubing to M-M columns. Adapters with #50 Ace-Thred fit Ace large columns.
Informacje prawne
Swagelok is a registered trademark of Swagelok Company
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)