Merck
Wszystkie zdjęcia(4)
Kluczowe dokumenty
Z181714
Supelco
Bottom-drip adapter for large columns
1/4-28 UNF 2B internal thread, for use with tubing connectors, Nylon
About This Item
Kod UNSPSC:
12000000
NACRES:
SB.52
Materiały
Nylon
producent / nazwa handlowa
Ace Glass 583814
metody
LPLC: suitable
Opis ogólny
Shallow taper of upper end minimizes mixing below the PE packing support disc (included). All adapters have an Ace Thred #50 and include one FETFE® O-ring.
Informacje prawne
FETFE is a registered trademark of Ace Glass, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)