Kluczowe dokumenty
Z180165
Supelco
End-fitting adapter for large columns
NPTF 1/2 in.
About This Item
Kod UNSPSC:
12352200
NACRES:
SB.52
producent / nazwa handlowa
Ace Glass 585720
metody
LPLC: suitable
gwint wewn.
1/2 in.
Opis ogólny
Polypropylene. Connects tubing adapters with NPT threads to column. Supplied with a locking retainer to hold 350 μm support screen (included) firmly in place. All adapters have an AceThred #50 and include one FETFE® O-ring
Informacje prawne
FETFE is a registered trademark of Ace Glass, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)