Merck
Z150320
Sigma-Aldrich
Aldrich® septum-inlet adapter, male
Plain, joint: ST/NS 24/40
About This Item
Kod UNSPSC:
41121815
NACRES:
NB.35
Połączenie
joint: ST/NS24/40
Opis ogólny
Inlet diam. 14 mm. Requires septum Z564702 or Z564699.
Septum-inlets require septum Z565695 or Z565679 except where noted.
Powiązanie
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informacje prawne
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC