Informacje o tej pozycji
NACRES:
NB.51
UNSPSC Code:
41121804
Capacity:
500 mL, 500 mL
Material:
borosilicate glass flask, round bottom , single-neck
Joint:
joint: ST/NS24/40
Packaging:
pkg of 1 ea
Pomoc techniczna
Pozwól nam pomócmaterial
borosilicate glass flask, round bottom , single-neck
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F304500
capacity
500 mL, 500 mL
General description
Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Still not finding the right product?
Explore all of our products under Synthware™ single-neck round-bottom flask
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Numer pozycji handlu globalnego
| SKU | NUMER GTIN |
|---|---|
| SYNF304500-1EA | 04061832208374 |