Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
1 ea | Estimated to ship onMay 26, 2026fromMILWAUKEE | $92.40 |
About This Item
NACRES:
NB.51
UNSPSC Code:
41121800
$92.40
Estimated to ship onMay 26, 2026Details
Subscriptions are currently unavailable.
Ships Every 4 weeks
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware C551190
jacket L
190 mm
General description
With narrow annular space for high cooling efficiency.
Legal Information
Synthware is a trademark of Kemtech America Inc.
1 of 1
This Item | |||
|---|---|---|---|
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 |
| manufacturer/tradename Synthware C551190 | manufacturer/tradename Synthware C554200 | manufacturer/tradename Synthware C154100 | manufacturer/tradename Synthware C554300 |
| jacket L 190 mm | jacket L 200 mm | jacket L 100 mm | jacket L 300 mm |
Still not finding the right product?
Explore all of our products under Synthware™ West condenser with removable hose connections
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.