Merck
R574635
N-(3,4-DIMETHYLPHENYL)-4-METHOXYBENZAMIDE
AldrichCPR
Wybierz wielkość
Informacje o cenach i dostępności nie są obecnie dostępne.
Informacje o tej pozycji
Wzór liniowy:
C16H17NO2
Numer CAS:
Masa cząsteczkowa:
255.319
MDL number:
Przejdź do
Pomoc techniczna
Pozwól nam pomóc
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)