Skip to Content
Merck
All Photos(1)

Key Documents

G9641

Sigma-Aldrich

Giemsa stain

powder, BioReagent, suitable for cell culture

Synonym(s):

Azure mixture sicc. Giemsa stain

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14ClN3S
CAS Number:
Molecular Weight:
291.80
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Pricing and availability is not currently available.

product line

BioReagent

form

powder

technique(s)

cell culture | mammalian: suitable

mp

300 °C (lit.)

solubility

ethanol: 1 mg/mL

SMILES string

[Cl-].CN(C)c1ccc2N=C3C=CC(=[NH2+])C=C3Sc2c1

InChI

1S/C14H13N3S.ClH/c1-17(2)10-4-6-12-14(8-10)18-13-7-9(15)3-5-11(13)16-12;/h3-8,15H,1-2H3;1H

InChI key

NALREUIWICQLPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used as a chromosome stain and to differentiate nuclear morphology of platelets, RBCs, WBCs, and other cell types.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xianhua Yin et al.
Applied and environmental microbiology, 75(18), 5779-5786 (2009-07-28)
O island 48 (OI-48) of Escherichia coli consists of three functional gene clusters that encode urease, tellurite resistance (Te(r)), and putative adhesins Iha and AIDA-1. The functions of these clusters in enterohemorrhagic E. coli (EHEC) O157:H7 infection are unknown. Deletion
Iset Medina Vera et al.
PLoS pathogens, 7(9), e1002197-e1002197 (2011-09-13)
Apicomplexans are obligate intracellular parasites that invade host cells by an active process leading to the formation of a non-fusogenic parasitophorous vacuole (PV) where the parasite replicates within the host cell. The rhomboid family of proteases cleaves substrates within their
Chun-Li Xiao et al.
BMC cancer, 11, 150-150 (2011-04-26)
Hepatocellular carcinoma (HCC) usually has a dismal prognosis because of its limited response to current pharmacotherapy and high metastatic rate. Sulfated oligosaccharide has been confirmed as having potent antitumor activities against solid tumors. Here, we explored the preclinical effects and
Mollie Jurewicz et al.
Diabetes, 59(12), 3139-3147 (2010-09-16)
A number of clinical trials are underway to test whether mesenchymal stem cells (MSCs) are effective in treating various diseases, including type 1 diabetes. Although this cell therapy holds great promise, the optimal source of MSCs has yet to be
Lee M Bergman et al.
Molecular and cellular biology, 29(16), 4539-4551 (2009-06-10)
CtBPs (CtBP1 and CtBP2) act in the nucleus as transcriptional corepressors and in the cytoplasm as regulators of Golgi apparatus fission. Studies in which the expression or function of CtBPs has been inhibited have independently identified roles for CtBPs in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service