Skip to Content
Merck
All Photos(1)

Key Documents

V000910

Sigma-Aldrich

Picric acid

p.a.

Synonym(s):

2,4,6-Trinitrophenol, PA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
Colour Index Number:
10305
Beilstein:
423400
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

p.a.

Quality Level

expl. lim.

0.01 %

mp

122-123 °C (dried material) (lit.)

SMILES string

Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H

InChI key

OXNIZHLAWKMVMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameSkull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Desen. Expl. 1

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giuliana Magri et al.
Nature immunology, 15(4), 354-364 (2014-02-25)
Innate lymphoid cells (ILCs) regulate stromal cells, epithelial cells and cells of the immune system, but their effect on B cells remains unclear. Here we identified RORγt(+) ILCs near the marginal zone (MZ), a splenic compartment that contains innate-like B

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service