Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
technique(s)
immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... RAG1(5896)
General description
Immunogen
Application
Epigenetics & Nuclear Function
Chromatin Biology
Immunocytochemistry Analysis: 1:100 dilution from a representative lot detected RAG-1 in Jurkat cells.
DyLight® is a registered trademark of Thermo Fisher Scientific.
Physical form
Preparation Note
Analysis Note
Immunoprecipitation Analysis: 10 µg of this antibody immunoprecipitated RAG-1 in 500 µg of HL-60 cell lysate. Immunopreciptated protein was then probed with 1 µg/mL of the same antibody in WB.
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source rabbit | biological source mouse | biological source mouse | biological source rabbit |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form affinity isolated antibody |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level - |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human, rat, mouse |
| technique(s) immunocytochemistry: suitable, western blot: suitable, immunoprecipitation (IP): suitable | technique(s) ELISA: suitable, immunocytochemistry: suitable, western blot: suitable | technique(s) immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable | technique(s) western blot: 1:500-1:1000 |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| ABE440 | 04053252918032 |



