Cell Imaging Instruments
Cellular imaging instruments can be used to evaluate, measure, analyze, and assay the state of cells and cell cultures in a variety of workflows. Digital cell imagers utilize algorithms to provide objective measurements of cell and cell culture status, eliminating user bias and streamlining processes for greater consistency. Live cell imaging systems enable the study of living cells using time-lapse microscopy, yielding critical insights into cell function and multi-parameter data for basic research and drug discovery. We offer cost-effective solutions to expand your lab's capacity for live and digital cell imaging.
Products
Millicell® DCI Digital Cell Imager
Monitor your cell cultures with ease with the Millicell® DCI Digital Cell Imager. From confluency to cell count and morphology, the Millicell® DCI automates cell culture measurements for quicker, more consistent culturing. This instrument is ideal for scientists and lab technicians responsible for the growth and maintenance of cell cultures for basic research, drug discovery, and bioprocess applications.
- Objective measurements
- Confluency determination
- Estimated cell counts
- Hemocytometer or in-vessel measurement
- Adherent cell, spheroid, and organoid cultures
- Optional web-based cloud service for data storage, data archiving, and growth trend analysis
CellASIC® ONIX2 Live Cell Imaging System
Convert your microscope to a live cell imaging system with the CellASIC® ONIX2 system. This microfluidic system uses high-quality, optically clear microfluidic plates and intuitive software to integrate with a broad range of inverted microscopes to allow continuous, high magnification observation of live cells as they react to their environment in real-time.
- Dynamic live cell imaging
- Compatible with any inverted microscope
- Small footprint
- User-friendly operation
- Automatically control flow rates, gas and temperature shifts, standing gradients, nutrient/drug additions, and media changes
- Optically clear microfluidic plates allow live-cell imaging for many applications
Related Product Resources
- Flyer: Millicell® DCI Overview
The Millicell® DCI Digital Cell Imager provides quick, objective determination of common cell culture parameters including confluency, cell count, and morphology.
- Brochure: CellASIC® ONIX2 Overview
Precision control of your cell culture environment for advanced live cell imaging & microscopy.