主要文件
Z709972
Sartorius pipette tips
volume range 10-1000 μL, standard, refill, non-sterile
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121600
NACRES:
NB.01
最大体积:
1 mL
体积范围:
10-1000 μL
无菌性:
non-sterile
特点 :
DNase free
RNase free
barrier: no
endotoxin free
包装:
pack of 96 ea (racked)
制造商/商品名称:
Sartorius 791002
物料
colorless
无菌性
non-sterile
特点
DNase free
RNase free
barrier: no
endotoxin free
包装
pack of 96 ea (racked)
制造商/商品名称
Sartorius 791002
最大体积
1 mL
体积范围
10-1000 μL
正在寻找类似产品? 访问 产品对比指南
一般描述
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
其他说明
Browse all pipette tips
历史批次信息供参考:
分析证书(COA)
访问文档库
相关内容
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
PFAS,也被称为“永久化学物质”,会在环境中永久性存在,威胁人类健康。我们提供PFAS定量的环境监测工具,助力研究人员、监管人员和实验室的PFAS检测。
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
查看所有结果