主要文件
Z741659
Sartorius pipette tips
volume range 0.1-10 μL, Standard, non-sterile, bulk, pack of 1000 ea
登录查看公司和协议定价
About This Item
UNSPSC代码:
41121600
NACRES:
NB.01
最大体积:
10 μL
体积范围:
0.1-10 μL
无菌性:
non-sterile
特点 :
barrier: no
包装:
pack of 1000 ea
制造商/商品名称:
Sartorius 790014
物料
colorless
无菌性
non-sterile
特点
barrier: no
包装
pack of 1000 ea
制造商/商品名称
Sartorius 790014
最大体积
10 μL
体积范围
0.1-10 μL
正在寻找类似产品? 访问 产品对比指南
一般描述
The non-filtered Optifit Tips are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment.
Optifit Tips are available in various packaging types for all your needs:
  • Single Tray racks – purity certified tip racks
  • Refill Packs - purity certified and ecological refill packages
  • Refill Towers - space-saving and ecological refill packages
  • Flexibulk boxes - space-saving, ecological and economical bulk boxes
  • Standard bulk – economical bulk boxes
While Optifit Tip trays are colour coded for easy matching with a corresponding, colour coded Sartorius pipette, they do also match with most other pipette due to their universal design.
Strict quality standards, ISO 9001 and ISO 13485, are followed from tip R&D to production and delivery. The purity certified tips are manufactured in ISO 8 classified clean room conditions to avoid any contaminations and are tested for DNase, RNase and endotoxins per each tip lot. Pre-sterilised tips are also available for the purest laboratory work. Sartorius offers a selection of pipette tips, extended and large volume tips, for special needs.
其他说明
Browse all pipette tips
历史批次信息供参考:
分析证书(COA)