主要文件
CLS33982B
Corning® PYREX® glass housing filter discs
coarse frit
别名:
lab equipment accessory
登录查看公司和协议定价
About This Item
UNSPSC代码:
40161507
NACRES:
NB.43
产品名称
Pyrex® fritted support base with 40/35 joint, for 47 mm microfiltration assembly, coarse frit
物料
glass housing
包装
pack of 1
制造商/商品名称
Corning
直径
47 mm
颈直径
33-41 mm
法律信息
PYREX is a registered trademark of Corning, Inc.
历史批次信息供参考:
分析证书(COA)