主要文件
C9056
Sigma-Aldrich
盖玻片
size 24 mm × 60 mm
登录查看公司和协议定价
About This Item
UNSPSC代码:
41122600
NACRES:
MA.01
描述
No. 1 thickness (0.13-0.16 mm)
包装
case of 10 1-oz pkgs
pkg of ~54 per ounce
尺寸
24 mm × 60 mm
正在寻找类似产品? 访问 产品对比指南
历史批次信息供参考:
分析证书(COA)
访问文档库
相关内容
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.
Three-dimensional (3D) printing of biological tissue is rapidly becoming an integral part of tissue engineering.