HGTC050SP
1 of 10
Millipore
XX2004703
垫圈,PTFE
XX1002535
垫圈,PTFE涂层
58066
PTFE 垫圈
Milli-Q
ZSQ200U0T0
Milli-Q® SQ 200超纯水系统
Sigma-Aldrich
HPA001667
Anti-PAPPA antibody produced in rabbit
Z243620
Aldrich clamp holder
265M-1
HHV-8 (13B10) Mouse Monoclonal Antibody
YY2004050
Cartridge Housing Gasket 2 in. Silicone TC
TANKSQ050
Milli-Q® SQ XL 50 L水箱
CR99F1006
Polygard-CR 筒式过滤器 10 in.100M µm F 号
silicone
SP1
0.5 cm
for use with Series 2000 Sanitary Cartridge Housings
Filter Housings
Filtration
Filtration for Bioprocessing
Sterile Filtration and Bioburden Control