Key Documents
Z682438
Ace vapor duct tube, for condenser "A" assembly
joint: ST/NS 29/32, L 160 mm, fits Büchi® models R-210/R-215
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121800
NACRES:
NB.43
接合處
joint: ST/NS29/32
製造商/商標名
Büchi equivalent, 46964
長度
160 mm
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
The vapor duct tube accessories are for Büchi® condenser "A" assemblies, and will fit either Büchi evaporator models R-210/R-215 or R-114/R-144.
Combi-clips: not included.
法律資訊
Büchi is a registered trademark of Büchi Labortechnik AG