Key Documents
Z549479
Sigma-Aldrich
Stopcock Adapter
straight glass stopcock, joint: ST/NS 14/20
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41121815
NACRES:
NB.35
材料
straight glass stopcock
接合處
joint: ST/NS14/20
正在寻找类似产品? 访问 产品对比指南
相关类别
一般說明
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.