Key Documents
Z222127
Bransonic® ultrasonic cleaner accessories
Perforated Tray, stainless steel, tank capacity 9.2 L
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41101500
NACRES:
NB.74
材料
stainless steel
包裝
pack of 1 ea
製造商/商標名
Branson 100-410-166
罐容量
9.2 L
正在寻找类似产品? 访问 产品对比指南
一般說明
Perforated stainless steel tray for use with Branson M series, model 5800 Utrasonic Bath.
法律資訊
Bransonic is a registered trademark of Branson Ultrasonics Corp.
从最新的版本中选择一种:
分析证书(COA)