主要文件
Z203750
Sigma-Aldrich
Aldrich® large-capacity cold trap
Inside finger for traps with ST/NS: 55/50 joint
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104812
NACRES:
NB.43
正在寻找类似产品? 访问 产品对比指南
相关类别
一般描述
With removable cold-fingers and a choice of 55/50 joint or 50 mm O-ring joint. Traps with O-ring joint include pinch clamp. Use with or without dry ice to condense and collect in trap. Dewar flask F9651 is recommended for use with 500 mL capacity cold traps.
法律信息
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
历史批次信息供参考:
分析证书(COA)