主要文件
Z171727
Sigma-Aldrich
Vacuum-jacketed filter funnel
200 mL, 50 mm frit, 25 to 50 μm porosity
登录查看公司和协议定价
About This Item
UNSPSC代码:
41104900
NACRES:
NB.43
制造商/商品名称
Aldrich
容量
200 mL
200 mL
圆盘直径
50 mm
过滤器直径
50 mm
孔径
25-50 μm
一般描述
200 mL, 50 mm frit, 25 to 50 μm porosity. Useful for low-temperature filtrations. 45/50F top joint, 24/40M bottom joint. Includes 45/50M to 24/40F reducing adapter for top joint.
历史批次信息供参考:
分析证书(COA)