Key Documents
HAM201100
Hamilton® Gastight® syringe, general use, 1700 series
model, 1725, AccuDil (AD) end, volume 250 μL
登录查看公司和协议定价
About This Item
分類程式碼代碼:
41122001
NACRES:
NB.03
材料
Borosilicate glass/PTFE
Gastight syringe
特點
autoclavable: no
model 1725, AccuDil (AD) end
製造商/商標名
Hamilton
參數
20-50 °C temp. range (68-122 °F)
500 psig pressure
容積
250 μL
正在寻找类似产品? 访问 产品对比指南
法律資訊
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.