主要文件
HAM7656-01
Hamilton® TLC syringe
model, 1710 RN, no needle, volume 100 μL
登录查看公司和协议定价
About This Item
UNSPSC代码:
42142600
NACRES:
NB.03
物料
Gastight syringe
特点
autoclavable
model 1710 RN, no needle
制造商/商品名称
Hamilton
参数
10-115 °C temp. range (50-239 °F)
1000 psig pressure
内径 × 外径
1.46 mm × 7.75 mm
体积
100 μL
正在寻找类似产品? 访问 产品对比指南
一般描述
The needles on Thin Layer Chromatography (TLC) syringes are coated from the point for 3/4” with a PTFE-coating. The coating reduces the surface tension between the needle and the liquid making it ideal for reproducible sample spotting. The TLC syringe is available as a cemented needle (SNTLC) or as a replacement needle to be installed into a standard removable needle (RN) syringe. To find the appropriate no needle syringe select general syringes and filter by termination and gauge. Choose the “RN” and “No Needle Included” options to see all syringes that can receive TLC needles. To find the appropriate TLC needle select the TLC option under “Needles” in the main product menu.
法律信息
Hamilton is a registered trademark of Hamilton Co.
历史批次信息供参考:
分析证书(COA)