Pestizide
Materialien
Analytische Säule
Produkt-Nr.
Beschreibung
Preisangaben
Vorsäulenkartusche
Produkt-Nr.
Beschreibung
Preisangaben
Zubehör
Produkt-Nr.
Beschreibung
Preisangaben
Wheaton Weithalsflasche
amber soda-lime glass bottle, capacity 1000 mL, white polypropylene cap, (PTFE faced LDPE liner), case of 12 eaCoupler for Legacy Guard Column Holder
PEEK, I.D. × O.D. × Overall L 0.010 in. × 1/16 in. × 1 in.Supelguard Vorsäulenhalter
Stand-alone, for use with Supelguard cartridges (2 cm L. x 2.1 to 4.0 mm I.D.)CONDITIONS
column
Ascentis C8, 15 cm x 4.6 mm I.D., 5 μm particles (581424-U)
mobile phase
[A] water; [B] acetonitrile
gradient
35 to 75% B in 13 min; held for 5 min
flow rate
1.0 mL/min
column temp.
35 °C
detector
UV, 254 nm
injection
100 μL
sample
as indicated in 2.2% 90:10 acetonitrile:acetone in DI water