주요 문서
57513-U
Millipore
Porozorb Cartridge
Porozorb 4004, size 4000 mL
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩6,319,200
₩6,319,200
예상 입고일2025년 5월 15일세부사항
크기 선택
보기 변경
1 EA
₩6,319,200
About This Item
UNSPSC 코드:
23151817
NACRES:
SB.52
₩6,319,200
예상 입고일2025년 5월 15일세부사항
Quality Level
크기
4000 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Porozorb cartridge is a ready-to-use column containing Amberlite XAD®-4 polymeric hydrophobic resin. It removes detergents effectively, as well as isolates and/or purifies pharmaceutical and biological compounds. This cartridge is suitable for removing phenols and other organics in aqueous solutions, and for recovering antibodies.
기타 정보
Not for clinical or diagnostic use. Due to shelf life limitations, the cartridges are made upon receipt of an order; expect a 2-4 week delay in shipping times. Determination of cartridge purity for a particular application is the purchaser′s responsibility. Equilibrate the cartridge with matrix buffer prior to use.
법적 정보
Amberlite is a trademark of DuPont de Nemours, Inc.
Porozorb is a trademark of Sigma-Aldrich Co. LLC
XAD is a registered trademark of The Dow Chemical Company or an affiliated company of Dow
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)