21031
₩145,145
출고 가능일2025년 3월 31일세부사항
1 of 10
Supelco
21033
Filter Cartridge
06335
Potassium Standard for ICP
18021
Sulfur Standard for ICP
00462
Sodium Standard for ICP
19051
Calcium Standard for ICP
Sigma-Aldrich
Z290432
Sigma-Aldrich® vacuum filtration assembly
1.00441
Nitric acid 65%
Millipore
PM2547050
PTFE Membrane Filter, 2.0 μm Pore Size
SLGSVR255F
Millex™ MCE syringe filter
1.00456
EPA IP-8,TO-10A
pkg of 1 ea
active air sampling: suitable
air monitoringenvironmentalindustrial hygiene
가장 최신 버전 중 하나를 선택하세요:
It looks like we've run into a problem, but you can still download Certificates of Analysis from our 문서 section.