주요 문서
Z740382
Eco-Friendly SpecTainer, biodegradable
size 60 mL, sterile
로그인조직 및 계약 가격 보기
크기 선택
500 EA
₩492,275
₩492,275
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
500 EA
₩492,275
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.22
₩492,275
구입 가능 여부는 고객센터에 문의하십시오.
material
polyethylene cap
polypropylene container
무균
sterile
포장
pack of 500 ea
제조업체/상표
Simport C567-60AQSECO
크기
60 mL
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Simport Eco-Friendly SpecTainer will biodegrade to become some of the soil′s organic components in less than 7 years in constant contact with the soil instead of up to 400 years when using conventional plastics.The Simport Eco-Friendly SpecTainer, one of the most innovative specimen containers on the market today, is now available in a new version: The Eco-Friendly SpecTainer, a rapidly degradable Specimen Container. Plastics take hundreds of years to degrade naturally in the environment. The only condition that is necessary for the Eco-Friendly SpecTainer to biodegrade is constant contact with other degrading material. No heat, physical stress, oxygen or sunlight necessary. The Eco-Friendly SpecTainer has the ability to break down safely and quickly, by biological means, into the raw materials of nature and disappear into the environment.Does not contain heavy metals. No negative effect to its physical properties. No heat, light or mechanical stress needed for product breakdown. No toxic residue. Fully degradable within 7 years, aerobically or anaerobically.
법적 정보
SpecTainer is a trademark of Simport
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)