주요 문서
Z380156
Sigma-Aldrich
Security bag for X-ray film
For 8 × 10 inch (20 × 25 cm) film
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
24111500
NACRES:
NB.31
포장
pack of 1 ea
제조업체/상표
Bel-Art F13352-0001
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Contains and protects a whole box of x-ray film in a light-tight environment. The fabric is tightly woven black nylon coated inside with opaque black vinyl. The overlapping flap closure secures the bag on all edges with a hook-and-loop seal.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Xiang Li et al.
Nucleic acids research, 47(10), 5074-5085 (2019-06-05)
In microorganisms, a number of metalloproteins including PerR are found to regulate gene expression in response to environmental reactive oxygen species (ROS) changes. However, discovery of similar regulatory mechanisms remains elusive within mammalian cells. As an antioxidant metalloenzyme that maintains