주요 문서
Z328847
BRAND® magnetic stirring bar, PTFE, cylindrical
L 50 mm, bar diam. 8 mm
로그인조직 및 계약 가격 보기
크기 선택
10 EA
₩104,342
₩104,342
출고 가능일2025년 4월 16일세부사항
크기 선택
보기 변경
10 EA
₩104,342
About This Item
UNSPSC 코드:
41103806
eCl@ss:
32100100
NACRES:
NB.72
₩104,342
출고 가능일2025년 4월 16일세부사항
material
PTFE-coated
cylindrical
제조업체/상표
BRAND 137135
길이
50 mm
막대 직경
8 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
특징 및 장점
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
기타 정보
BR137135 (India)
법적 정보
BRAND is a registered trademark of BRAND GMBH + CO KG
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)