GBL470635
Grace Bio-Labs ProPlate® microarray system, slide module only
well size 3, L × W × depth 20 mm × 20 mm × 7.6 mm, without clips
동의어(들):
incubation chambers, microarray hybridization chambers, slide chambers
로그인조직 및 계약 가격 보기
크기 선택
10 EA
₩245,732
₩245,732
예상 입고일2025년 5월 20일세부사항
모든 사진(1)
크기 선택
보기 변경
10 EA
₩245,732
About This Item
UNSPSC 코드:
41121800
NACRES:
NB.17
₩245,732
예상 입고일2025년 5월 20일세부사항
추천 제품
material
black silicone (on Delrin)
특징
without clips
포장
pack of 10 ea
제조업체/상표
Grace Bio-Labs 470635
길이 × 너비 × 깊이
20 mm × 20 mm × 7.6 mm
웰 크기
3
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Grace Bio-Labs ProPlate® multi-array chambers (microarray systems) are designed to be used with ONCYTE® nitrocellulose-coated slides. These multi-array chambers integrate microscope slide based microarray technology with high-throughput (HTS applications) microtiter plate processing. Up to 64 individual arrays can be assayed per slide.
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.
Features:
- Up to 64 leak-proof wells on any slide surface without the use of adhesive
- Microtiter plate well spacing facilitates use with multichannel pipettes
- 7 x 7 mm square wells are ideal for high content arrays 5-350 ul well volumes accommodate small reagent and large wash volumes
법적 정보
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.
가장 최신 버전 중 하나를 선택하세요:
활성 필터