Merck
MCE2513FS1
Millipore
Millistak+® Pod Depth Filter
CE25, filtration area 1.4m², Process Scale
로그인조직 및 계약 가격 보기
About This Item
UNSPSC 코드:
23151806
가격 및 재고 정보를 현재 이용할 수 없음
제품명
Millistak+® Pod Depth Filter, CE25 (media series), cellulose membrane, effective filtration area 1.4 m2
material
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane
polypropylene housing (glass-filled)
Quality Level
멸균 적합성
autoclavable compatible
유효 여과 면적
1.4 m2
피팅
seal (flat seal)
카트리지 코드
CE25 (media series)
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MCE2513FS1 comes in a POD format.
This CE25 media series delivers optimal performance through a gradient density matri, ideal for primary (coarse) clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
특징 및 장점
  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces
법적 정보
MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10-13 - German Storage Class 10 to 13
Emprove® 프로그램이란?
Emprove® 프로그램은 자사의 필터 그리고 일회용 구성품, 제약용 원료 물질 및 시작 물질의 종합적이고 빈틈없는 문건을 제공하는 체계입니다.
Material Qualification Dossier
무료 - 다운로드하려면 로그인하십시오
혜택 자세히 보기 Emprove®프로그램 구독권은 다음을 제공합니다
이러한 문서를 이용할 수 없는 이유는?
Emprove® 자료는 제약 생산자의 등록 과정 및 위험성 평가 활동을 지원하도록 의도된 정보 그리고 기밀 정보를 포함합니다. 열람하기 원하시면 문의 사항 하십시오.