주요 문서
Z266280
Sigma-Aldrich
Polymer-body combination pH electrodes
BNC fitting
로그인조직 및 계약 가격 보기
크기 선택
1 EA
₩467,306
₩467,306
예상 입고일2025년 4월 21일세부사항
크기 선택
보기 변경
1 EA
₩467,306
About This Item
UNSPSC 코드:
41115603
NACRES:
NB.79
₩467,306
예상 입고일2025년 4월 21일세부사항
스템 길이 × 직경
183 mm × 6 mm
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Sealed, gel-filled
Long, narrow/tubes, vials.
The combination of a pH indicating and reference half-cell in an impact-resistant epoxy body. Designed for applications where breakage is a problem.
  • Environmental - field sampling
  • Industrial - process control
  • Educational - core courses
Rugged bulb, fast response in optimal range of 0 to 12 pH. Suggested temp. range: 0 to 80 °C. For Tris, sulfides, and heavy metals, choose a double junction reference electrode. Supplied with 36 in. lead.
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)