주요 문서
Z245240
ACE microscale glassware kit
MICRO/MINI-Lab® Basic kit II
로그인조직 및 계약 가격 보기
크기 선택
1 KIT
₩1,044,131
₩1,044,131
구입 가능 여부는 고객센터에 문의하십시오.
크기 선택
보기 변경
1 KIT
₩1,044,131
About This Item
UNSPSC 코드:
41100000
NACRES:
NB.43
₩1,044,131
구입 가능 여부는 고객센터에 문의하십시오.
제조업체/상표
Ace Glass 9560-30
유사한 제품을 찾으십니까? 방문 제품 비교 안내
일반 설명
All joints threaded 14/10, supplied in a clear, flexible plastic box with tilt lid. Includes magnetic "V" stirrer (not shown).
Designed to give students experience with a wide range of organic reactions with increased safety. Kits contain microscale glassware needed to perform experiments described in the textbook Microscale Organic Laboratory with Multistep and Multiscale Syntheses.
Greaseless, threaded ST14/10 connections can be interfaced with traditional ST14/10 joints. Conical reaction vials include open-top caps, septa, and O-rings. Glassware in the DELUXE kit is supplied in a foam-lined plastic carrying case. The BASIC kit is supplied in a clear plastic, compartmentalized box with tilt lid.
법적 정보
Micro/Mini-Lab is a registered trademark of Ace Glass, Inc.
관련 제품
제품 번호
설명
가격
가장 최신 버전 중 하나를 선택하세요:
시험 성적서(COA)
문서 라이브러리 방문
Ze-Quan Mo et al.
Fish & shellfish immunology, 47(1), 280-288 (2015-09-20)
CCR6 have been demonstrated playing an important role in immune cells homing to mucosal tissues, mediating antigen presentation and immune response in mammals. CCR6 in lower vertebrate leukocyte homing has not yet been revealed. Cryptocaryon irritans is believed to be
Xi Chen et al.
Immunobiology, 221(1), 48-55 (2015-08-27)
Macrophages function as an essential component of innate immune system, contributing to both the initiation and appropriate resolution of inflammation. The exposure of macrophages to the microbial products, such as lipopolysaccharide (LPS), can strongly shift the balance between tissue homeostasis
Xia Pan et al.
Food chemistry, 188, 569-575 (2015-06-05)
Polygalacturonase is one of the most important industrial pectinases. To enrich the genetic resources and develop new enzyme candidates, three polygalacturonase genes (Nfpg I-III) of glycosyl hydrolase family 28 were cloned from Neosartorya fischeri P1 and functionally expressed in Pichia